Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TCF20
Entrez Gene ID
523151
Gene Name
transcription factor 20
Gene Aliases
-
Chromosome Location
Chr.5: 113101868 - 113270674 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192574.2 NP_001179503.2 4 - 5 5951 61

View Full Product Details