Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CRLF3
Entrez Gene ID
528033
Gene Name
cytokine receptor like factor 3
Gene Aliases
-
Chromosome Location
Chr.19: 18107484 - 18149013 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192650.1 NP_001179579.1 3 - 4 457 66

View Full Product Details