Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CLDND2
Entrez Gene ID
618259
Gene Name
claudin domain containing 2
Gene Aliases
-
Chromosome Location
Chr.18: 57436427 - 57438684 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206293.1 NP_001193222.1 2 - 3 308 54

View Full Product Details