Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLAMF6
Entrez Gene ID
508183
Gene Name
SLAM family member 6
Gene Aliases
-
Chromosome Location
Chr.3: 9152808 - 9175511 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206364.1 NP_001193293.1 6 - 7 888 102
GenBank mRNA BC102832.1 - 6 - 7 875 102

View Full Product Details