Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NDUFAF5
Entrez Gene ID
522230
Gene Name
NADH:ubiquinone oxidoreductase complex assembly factor 5
Gene Aliases
C13H20orf7
Chromosome Location
Chr.13: 7387507 - 7412144 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206395.1 NP_001193324.1 10 - 11 981 89
GenBank mRNA BC122720.1 - 10 - 11 982 89

View Full Product Details