Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LANCL3
Entrez Gene ID
519239
Gene Name
LanC like 3
Gene Aliases
-
Chromosome Location
Chr.X: 105734791 - 105847786 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206408.1 NP_001193337.1 3 - 4 1214 58

View Full Product Details