Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
CEP83
Entrez Gene ID
618145
Gene Name
centrosomal protein 83
Gene Aliases
CCDC41
Chromosome Location
Chr.5: 24070150 - 24216286 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206525.1 NP_001193454.1 8 - 9 1296 112

View Full Product Details