Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FOLR3
Entrez Gene ID
516067
Gene Name
folate receptor 3
Gene Aliases
FBP, FR-alpha
Chromosome Location
Chr.15: 51782877 - 51788148 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206527.1 NP_001193456.1 1 - 2 120 75
GenBank mRNA DT858714.1 - 1 - 2 56 75
DY099458.1 - 1 - 2 120 75

View Full Product Details