Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TAS1R2
Entrez Gene ID
529847
Gene Name
taste 1 receptor member 2
Gene Aliases
-
Chromosome Location
Chr.2: 133800882 - 133813306 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206529.3 NP_001193458.3 1 - 2 176 67

View Full Product Details