Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PLEKHS1
Entrez Gene ID
529092
Gene Name
pleckstrin homology domain containing S1
Gene Aliases
C26H10orf81
Chromosome Location
Chr.26: 34215281 - 34242464 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206558.1 NP_001193487.1 8 - 9 786 84

View Full Product Details