Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PPIL3
Entrez Gene ID
615703
Gene Name
peptidylprolyl isomerase like 3
Gene Aliases
-
Chromosome Location
Chr.2: 89518973 - 89530587 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206564.1 NP_001193493.1 1 - 2 70 107
GenBank mRNA BC110167.1 - 1 - 2 336 107
DV869571.1 - 1 - 2 56 107
EH160667.1 - 1 - 2 343 107

View Full Product Details