Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HCN1
Entrez Gene ID
538255
Gene Name
hyperpolarization activated cyclic nucleotide gated potassium channel 1
Gene Aliases
-
Chromosome Location
Chr.20: 29111349 - 29562326 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206580.2 NP_001193509.1 1 - 2 408 104

View Full Product Details