Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
USH1G
Entrez Gene ID
531104
Gene Name
USH1 protein network component sans
Gene Aliases
-
Chromosome Location
Chr.19: 56501076 - 56507949 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192702.3 NP_001179631.1 1 - 2 346 57

View Full Product Details