Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PI15
Entrez Gene ID
532863
Gene Name
peptidase inhibitor 15
Gene Aliases
-
Chromosome Location
Chr.14: 38154334 - 38191011 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192742.1 NP_001179671.1 2 - 3 435 68

View Full Product Details