Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GABRB2
Entrez Gene ID
540657
Gene Name
gamma-aminobutyric acid type A receptor subunit beta2
Gene Aliases
-
Chromosome Location
Chr.7: 72979171 - 73272335 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192760.2 NP_001179689.2 8 - 9 846 97

View Full Product Details