Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IKZF4
Entrez Gene ID
540671
Gene Name
IKAROS family zinc finger 4
Gene Aliases
ZNFN1A4
Chromosome Location
Chr.5: 57289459 - 57314348 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192764.1 NP_001179693.1 4 - 5 548 92

View Full Product Details