Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CSPG4
Entrez Gene ID
533653
Gene Name
chondroitin sulfate proteoglycan 4
Gene Aliases
-
Chromosome Location
Chr.21: 33182725 - 33218500 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192782.3 NP_001179711.2 8 - 9 4938 65

View Full Product Details