Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EEF2KMT
Entrez Gene ID
531984
Gene Name
eukaryotic elongation factor 2 lysine methyltransferase
Gene Aliases
FAM86A, eEF2-KMT
Chromosome Location
Chr.25: 4101434 - 4112376 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163938.1 NP_001157410.1 7 - 8 934 71
GenBank mRNA BT025354.1 - 7 - 8 911 71

View Full Product Details