Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DUSP13
Entrez Gene ID
616048
Gene Name
dual specificity phosphatase 13
Gene Aliases
-
Chromosome Location
Chr.28: 30868139 - 30881163 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163944.2 NP_001157416.1 4 - 5 577 89
GenBank mRNA BC111275.1 - 4 - 5 578 89

View Full Product Details