Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CELA3B
Entrez Gene ID
281752
Gene Name
chymotrypsin-like elastase family, member 3B
Gene Aliases
ELA3B
Chromosome Location
Chr.2: 130816944 - 130824150 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166518.3 NP_001159990.3 6 - 7 664 88
GenBank mRNA AY057840.1 - 6 - 7 643 88

View Full Product Details