Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
PAK2
Entrez Gene ID
513673
Gene Name
p21 (RAC1) activated kinase 2
Gene Aliases
-
Chromosome Location
Chr.1: 71257814 - 71350400 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206727.1 NP_001193656.1 5 - 6 530 95

View Full Product Details