Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MTFMT
Entrez Gene ID
286855
Gene Name
mitochondrial methionyl-tRNA formyltransferase
Gene Aliases
-
Chromosome Location
Chr.10: 11936564 - 11961486 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166523.3 NP_001159995.1 1 - 2 234 84
GenBank mRNA AB004316.1 - 1 - 2 169 84

View Full Product Details