Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PTGES2
Entrez Gene ID
493639
Gene Name
prostaglandin E synthase 2
Gene Aliases
MPGES-2
Chromosome Location
Chr.11: 98758686 - 98764051 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166554.1 NP_001160026.1 3 - 4 532 67

View Full Product Details