Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HSPD1
Entrez Gene ID
511913
Gene Name
heat shock protein family D (Hsp60) member 1
Gene Aliases
HSP60
Chromosome Location
Chr.2: 86035475 - 86045870 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166608.1 NP_001160080.1 10 - 11 1744 106
NM_001166609.1 NP_001160081.1 10 - 11 1629 106
NM_001166610.1 NP_001160082.1 10 - 11 1496 106

View Full Product Details