Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AQP5
Entrez Gene ID
782368
Gene Name
aquaporin 5
Gene Aliases
-
Chromosome Location
Chr.5: 29908595 - 29913170 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191160.1 NP_001178089.1 1 - 2 568 69

View Full Product Details