Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SAMD12
Entrez Gene ID
783254
Gene Name
sterile alpha motif domain containing 12
Gene Aliases
-
Chromosome Location
Chr.14: 45625075 - 46074096 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191180.2 NP_001178109.1 3 - 4 458 123

View Full Product Details