Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCND2
Entrez Gene ID
540416
Gene Name
potassium voltage-gated channel subfamily D member 2
Gene Aliases
-
Chromosome Location
Chr.4: 84688361 - 85252121 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191184.1 NP_001178113.1 5 - 6 1720 74

View Full Product Details