Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ELK1
Entrez Gene ID
786886
Gene Name
ETS transcription factor ELK1
Gene Aliases
-
Chromosome Location
Chr.X: 86007074 - 86022529 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191236.1 NP_001178165.1 5 - 6 1343 114

View Full Product Details