Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SGO2
Entrez Gene ID
532412
Gene Name
shugoshin 2
Gene Aliases
SGOL2
Chromosome Location
Chr.2: 89070468 - 89099327 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191290.3 NP_001178219.2 8 - 9 4070 118
GenBank mRNA EV738882.1 - 1 - 2 374 118

View Full Product Details