Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
BMP8A
Entrez Gene ID
788948
Gene Name
bone morphogenetic protein 8a
Gene Aliases
-
Chromosome Location
Chr.3: 106544060 - 106584726 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206869.2 NP_001193798.2 3 - 4 681 53

View Full Product Details