Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NTN5
Entrez Gene ID
785970
Gene Name
netrin 5
Gene Aliases
-
Chromosome Location
Chr.18: 55294153 - 55300421 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206872.1 NP_001193801.1 4 - 5 1021 91
GenBank mRNA EV739345.1 - 2 - 3 729 91

View Full Product Details