Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TSPAN32
Entrez Gene ID
530507
Gene Name
tetraspanin 32
Gene Aliases
-
Chromosome Location
Chr.29: 49261072 - 49284265 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242955.2 NP_001229884.1 7 - 8 633 75

View Full Product Details