Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SYNE4
Entrez Gene ID
509061
Gene Name
spectrin repeat containing nuclear envelope family member 4
Gene Aliases
C18H19orf46, C19orf46
Chromosome Location
Chr.18: 46675916 - 46680973 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243287.1 NP_001230216.1 1 - 2 222 82
GenBank mRNA BC149244.1 - 1 - 2 223 82

View Full Product Details