Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GALNTL6
Entrez Gene ID
615819
Gene Name
polypeptide N-acetylgalactosaminyltransferase like 6
Gene Aliases
-
Chromosome Location
Chr.8: 3940092 - 5482560 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191298.2 NP_001178227.1 8 - 9 1148 103

View Full Product Details