Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPP1R1C
Entrez Gene ID
784202
Gene Name
protein phosphatase 1 regulatory inhibitor subunit 1C
Gene Aliases
-
Chromosome Location
Chr.2: 14473125 - 14594168 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191352.1 NP_001178281.1 2 - 3 139 76

View Full Product Details