Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WHAMM
Entrez Gene ID
510810
Gene Name
WASP homolog associated with actin, golgi membranes and microtubules
Gene Aliases
-
Chromosome Location
Chr.21: 23081878 - 23100087 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191456.1 NP_001178385.1 1 - 2 705 113

View Full Product Details