Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PIDD1
Entrez Gene ID
100137737
Gene Name
p53-induced death domain protein 1
Gene Aliases
LRDD, PIDD
Chromosome Location
Chr.29: 50560866 - 50567587 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191487.1 NP_001178416.1 12 - 13 2170 97

View Full Product Details