Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ELF4
Entrez Gene ID
504514
Gene Name
E74 like ETS transcription factor 4
Gene Aliases
-
Chromosome Location
Chr.X: 14042900 - 14081548 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192029.1 NP_001178958.1 3 - 4 338 76
GenBank mRNA KU695668.1 - 3 - 4 338 76

View Full Product Details