Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HEY2
Entrez Gene ID
538619
Gene Name
hes related family bHLH transcription factor with YRPW motif 2
Gene Aliases
-
Chromosome Location
Chr.9: 25546012 - 25561094 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192055.1 NP_001178984.1 2 - 3 163 108

View Full Product Details