Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DIRAS2
Entrez Gene ID
538667
Gene Name
DIRAS family GTPase 2
Gene Aliases
-
Chromosome Location
Chr.8: 87226438 - 87262862 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192064.3 NP_001178993.1 1 - 2 62 79

View Full Product Details