Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCN2B
Entrez Gene ID
538722
Gene Name
sodium voltage-gated channel beta subunit 2
Gene Aliases
-
Chromosome Location
Chr.15: 28855749 - 28871361 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192074.2 NP_001179003.1 1 - 2 167 73

View Full Product Details