Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLRN1
Entrez Gene ID
538836
Gene Name
clarin 1
Gene Aliases
-
Chromosome Location
Chr.1: 117161151 - 117204819 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192101.1 NP_001179030.1 2 - 3 434 97

View Full Product Details