Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDX1
Entrez Gene ID
538927
Gene Name
pancreatic and duodenal homeobox 1
Gene Aliases
-
Chromosome Location
Chr.12: 32118971 - 32123944 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192136.1 NP_001179065.1 1 - 2 412 72

View Full Product Details