Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FLVCR2
Entrez Gene ID
507318
Gene Name
feline leukemia virus subgroup C cellular receptor family, member 2
Gene Aliases
-
Chromosome Location
Chr.10: 86763436 - 86818617 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192143.1 NP_001179072.1 9 - 10 1449 77

View Full Product Details