Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SSC4D
Entrez Gene ID
539182
Gene Name
scavenger receptor cysteine rich family member with 4 domains
Gene Aliases
SRCRB4D
Chromosome Location
Chr.25: 34415457 - 34428404 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192192.2 NP_001179121.2 5 - 6 908 106

View Full Product Details