Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SSC4D
Entrez Gene ID
539182
Gene Name
scavenger receptor cysteine rich family member with 4 domains
Gene Aliases
SRCRB4D
Chromosome Location
Chr.25: 34415457 - 34428404 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192192.2 NP_001179121.2 7 - 8 1058 97

View Full Product Details