Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PODN
Entrez Gene ID
509606
Gene Name
podocan
Gene Aliases
-
Chromosome Location
Chr.3: 93157910 - 93183254 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192227.1 NP_001179156.1 1 - 2 41 54

View Full Product Details