Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TNFAIP1
Entrez Gene ID
539674
Gene Name
TNF alpha induced protein 1
Gene Aliases
-
Chromosome Location
Chr.19: 19822118 - 19832329 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192290.1 NP_001179219.1 3 - 4 615 100

View Full Product Details