Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDK6
Entrez Gene ID
511754
Gene Name
cyclin dependent kinase 6
Gene Aliases
-
Chromosome Location
Chr.4: 9929529 - 10190016 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192301.2 NP_001179230.1 2 - 3 527 78

View Full Product Details