Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NIPAL1
Entrez Gene ID
539757
Gene Name
NIPA like domain containing 1
Gene Aliases
NPAL1
Chromosome Location
Chr.6: 66717013 - 66744153 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192302.1 NP_001179231.1 1 - 2 115 103

View Full Product Details